Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Diethylene glycol monobutyl ether
Synonym(s):
2-(2-Butoxyethoxy)ethanol, BDG, Butyldiglycol, Diethylene glycol monobutyl ether
Linear Formula:
CH
3
(CH
2
)
3
OCH
2
CH
2
OCH
2
CH
2
OH
CAS Number:
112-34-5
Molecular Weight:
162.23
EC Number:
203-961-6
Products
Product Category
building blocks (1)
solvents (1)
Brand
Sigma-Aldrich (2)
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
32250
Diethylene glycol monobutyl ether,
≥98.0% (GC)
Expand
View Pricing
8.03129
Diethylene glycol monobutyl ether,
for synthesis
Expand
View Pricing
03351
Diethylene glycol monobutyl ether,
for surfactant analysis, ≥99.0%
Expand
View Pricing