
Diclofenac potassium

Synonym(s):
Diclofenac potassium salt, 2-[(2,6-Dichlorophenyl)amino]benzeneacetic acid potassium salt
Empirical Formula (Hill Notation):
C14H10Cl2KNO2
CAS Number:
Molecular Weight:
334.24
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3