
Dichlorvos

Synonym(s):
2,2-Dichlorovinyl dimethyl phosphate, DDVP
Empirical Formula (Hill Notation):
C4H7Cl2O4P
CAS Number:
Molecular Weight:
220.98
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing