Dibromoborane dimethyl sulfide complex solution
Synonym(s):
Dibromo(dimethyl sulfide)(hydro)boron solution, Dibromo(dimethyl sulfide)borane solution, Dibromoborane compound with dimethylsulfane solution, Dibromoborane dimethyl sulfide solution
Linear Formula:
(CH3)2S · BHBr2
CAS Number:
Molecular Weight:
233.76
Products
Brand
Formula Weight
Physical Form
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing