Skip to Content
Merck
Dibenzo-21-crown-7 97%

Dibenzo-21-crown-7

Dibenzo-21-crown-7 97%
Empirical Formula (Hill Notation):
C22H28O7
CAS Number:
Molecular Weight:
404.45

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing