Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Diamide
Synonym(s):
1,1′-Azobis(
N
,
N
-dimethylformamide),
N
,
N
,
N
′,
N
′-Tetramethylazodicarboxamide, Azodicarboxylic acid bis(dimethylamide), Diazenedicarboxylic acid bis(
N
,
N
-dimethylamide), TMAD
Linear Formula:
(CH
3
)
2
NCON=NCON(CH
3
)
2
CAS Number:
10465-78-8
Molecular Weight:
172.19
EC Number:
233-951-7
Products
Brand
Sigma-Aldrich (1)
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
D3648
Diamide,
Expand
View Pricing