Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
(+)-γ-Tocopherol
Synonym(s):
(
R
,
R
,
R
)-γ-Tocopherol, 7,8-Dimethyltocol
Empirical Formula (Hill Notation):
C
28
H
48
O
2
CAS Number:
54-28-4
Molecular Weight:
416.68
EC Number:
200-201-5
Products
Product Category
vitamins (2)
Brand
Sigma-Aldrich (1)
Supelco (1)
Color
Formula Weight
Gene Alias
Physical Form
Purity
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
T1782
(+)-γ-Tocopherol,
≥96% (HPLC)
Expand
View Pricing
47785
(+)-γ-Tocopherol,
analytical standard
Expand
View Pricing