Skip to Content
Merck
Deuterium oxide-18O 5 atom % D, 5 atom % 18O

Deuterium oxide-18O

Deuterium oxide-18O 5 atom % D, 5 atom % 18O
Synonym(s):
Doubly Labeled Water, Heavy water-d2,18O, Unnormalized water-18O
Linear Formula:
D218O
CAS Number:
Molecular Weight:
22.03

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Quality Segment
Isotope