Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Density Standard 749 kg/m
3
Synonym(s):
Dodecane
Linear Formula:
CH
3
(CH
2
)
10
CH
3
CAS Number:
112-40-3
Molecular Weight:
170.33
EC Number:
203-967-9
Products
Brand
Supelco (1)
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
76081
Density Standard 749 kg/m
3
,
H&D Fitzgerald Ltd. Quality
Expand
View Pricing