
D-Camphor sulfonic acid

Synonym(s):
(+)-Camphor-10-sulfonic acid, (+)-β-Camphorsulfonic acid, (1S)-(+)-Camphor-10-sulfonic acid
Empirical Formula (Hill Notation):
C10H16O4S
CAS Number:
Molecular Weight:
232.30
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
Optical Isomer
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing