
D−(−)-α-Phenylglycine

Synonym(s):
(R)-(−)-2-Phenylglycine, D-2-Phenylglycine, R-(−)-α-Aminophenylacetic acid, (2R)-2-Amino-2-phenylacetic acid, (R)-(−)-α-Aminophenylacetic acid
Linear Formula:
C6H5CH(NH2)CO2H
CAS Number:
Molecular Weight:
151.16
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2