Skip to Content
Merck
Cytisine phyproof® Reference Substance

Cytisine

Cytisine phyproof® Reference Substance
Synonym(s):
(−)-Cytisine, (1R,5S)-1,2,3,4,5,6-Hexahydro-1,5-methano-8H-pyrido[1,2a][1,5]diazocin-8-one, (1S,9S)-3,11-Diazatricyclo[7.3.1.03,8]trideca-5,7-dien-4-one, Baptitoxin, Laburnin, Sophorine, Ulexine
Empirical Formula (Hill Notation):
C11H14N2O
CAS Number:
Molecular Weight:
190.24
EC Number:

Products

Product Category
Boiling Point (°C)
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Research Area
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing