Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cyclophosphamide monohydrate
Synonym(s):
2-[Bis(2-chloroethyl)amino]tetrahydro-2
H
-1,3,2-oxazaphosphorine 2-oxide, Cytoxan
Empirical Formula (Hill Notation):
C
7
H
15
Cl
2
N
2
O
2
P · H
2
O
CAS Number:
6055-19-2
Molecular Weight:
279.10
EC Number:
200-015-4
Products
Product Category
bioactive small molecules (1)
Brand
Sigma-Aldrich (2)
Supelco (1)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Application
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
C0768
Cyclophosphamide monohydrate,
bulk package
Expand
View Pricing
C7397
Cyclophosphamide monohydrate,
ISOPAC
®
Expand
View Pricing
93813
Cyclophosphamide monohydrate,
analytical standard
Expand
View Pricing