
Cyclopentyl methyl ether

Synonym(s):
CPME
Empirical Formula (Hill Notation):
C6H12O
CAS Number:
Molecular Weight:
100.16
Products
Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Special Grade
Showing 1-4 of 4
Compare
Product Number
Description
Pricing