Skip to Content
Merck
Cyclohexanone uniLAB™

Cyclohexanone

Cyclohexanone uniLAB™
Synonym(s):
Cyclohexanone
Linear Formula:
C6H10(=O)
CAS Number:
Molecular Weight:
98.14

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Package Size
Reference Material Type
Special Grade
Technique