
Cyclohexane

Synonym(s):
Cyclohexane
Empirical Formula (Hill Notation):
C6H12
CAS Number:
Molecular Weight:
84.16
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-17 of 17
Compare
Product Number
Description
Pricing