
Copper(II) nitrate trihydrate

Synonym(s):
Cupric nitrate trihydrate, Copper(II) nitrate trihydrate
Linear Formula:
Cu(NO3)2 · 3H2O
CAS Number:
Molecular Weight:
241.60
Products
Product Category
Brand
Feature
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Special Grade