Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Conductivity water ( nominal 0 mS/cm)
Synonym(s):
Water
Linear Formula:
H
2
O
CAS Number:
7732-18-5
Molecular Weight:
18.02
Products
Brand
Supelco (1)
Boiling Point (°C)
Formula Weight
Physical Form
Quality Segment
Reference Material Type
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
1.01810
Conductivity water ( nominal 0 mS/cm),
Test solution for measurement of electrolytic conductivity, traceable to PTB Certipur
®
Expand
View Pricing