Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Conalbumin from chicken egg white
Synonym(s):
Ovotransferrin
CAS Number:
1391-06-6
EC Number:
215-727-0
Products
Product Category
proteins (2)
albumins (1)
media supplements (1)
Brand
Sigma-Aldrich (3)
Biological Source
Physical Form
Product Line
Tissue
Quality Segment
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
C7786
Conalbumin from chicken egg white,
BioReagent, suitable for cell culture
Expand
View Pricing
C0755
Conalbumin from chicken egg white,
Substantially iron-free
Expand
View Pricing
C0880
Conalbumin from chicken egg white,
Iron complex
Expand
View Pricing