Skip to Content
Merck
cis-4-Decenal technical grade, 80%

cis-4-Decenal

cis-4-Decenal technical grade, 80%
Linear Formula:
CH3(CH2)4CH=CHCH2CH2CHO
CAS Number:
Molecular Weight:
154.25
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing