
Ciprofloxacin Ethylenediamine Analog

Synonym(s):
7-[(2-Aminoethyl)amino]-1-cyclopropyl-6-fluoro-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid hydrochloride
Empirical Formula (Hill Notation):
C15H16FN3O3 · HCl
CAS Number:
Molecular Weight:
341.77
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing