Cholin acetate
Synonym(s):
2-Hydroxyethyl-trimethylammonium acetate, COL Acetate
Empirical Formula (Hill Notation):
C7H17NO3
CAS Number:
Molecular Weight:
163.21
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing