Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cholesteryl oleate
Synonym(s):
3β-Hydroxy-5-cholestene 3-oleate, 5-Cholesten-3β-ol 3-oleate, Cholesteryl cis-9-octadecenoate, Oleic acid cholesteryl ester
Empirical Formula (Hill Notation):
C
45
H
78
O
2
CAS Number:
303-43-5
Molecular Weight:
651.10
EC Number:
206-142-1
Products
Product Category
lipids (1)
metabolites (1)
steroids (1)
Brand
Sigma-Aldrich (1)
Biological Source
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
C9253
Cholesteryl oleate,
≥98% (HPLC; detection at 205 nm)
Expand
View Pricing