
Cholecalciferol Impurity B

Synonym(s):
7-Dehydrocholesterol, (−)-7-Dehydrocholesterol, 3β-Hydroxy-5,7-cholestadiene, 5,7-Cholestadien-3β-ol, Provitamin D3
Empirical Formula (Hill Notation):
C27H44O
CAS Number:
Molecular Weight:
384.64
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing