Skip to Content
Merck
Chloroform univAR™

Chloroform

Chloroform univAR™
Synonym(s):
Methylidyne trichloride, Trichloromethane, Chloroform
Empirical Formula (Hill Notation):
CHCl3
CAS Number:
Molecular Weight:
119.38

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Package Size
Special Grade
Technique
Showing 1-20 of 25
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2