
Chiralyst P618

Synonym(s):
Pentamethylcyclopentadienylrhodium(III) chloride dimer, 1,3-Cyclopentadiene, 1,2,3,4,5-pentamethyl-, rhodium complex, Bis(dichloro(pentamethylcyclopentadienyl)rhodium), Bis(pentamethylcyclopentadienylrhodium dichloride), Dichloro(pentamethylcyclopentadienyl)rhodium(II) dimer, Tetrachlorobis(pentamethylcyclopentadienyl)dirhodium
Linear Formula:
[Rh(C5Me5)Cl2]2
CAS Number:
Molecular Weight:
618.08
Products
Brand
Formula Weight
Physical Form
Quality Segment
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing