Cetirizine Related Compound C
Synonym(s):
2-(2-{4-[(4-Chlorophenyl)phenylmethyl]piperazin-1-yl}ethoxy)acetamide
Empirical Formula (Hill Notation):
C21H26ClN3O2
CAS Number:
Molecular Weight:
387.90
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing