Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cerium(III) iodide
Synonym(s):
Cerous triiodide
Linear Formula:
CeI
3
CAS Number:
7790-87-6
Molecular Weight:
520.83
EC Number:
232-228-3
Products
Brand
Sigma-Aldrich (1)
Formula Weight
Melting Point (°C)
Product Line
Purity
Quality Segment
Reagent Type
Core
Special Grade
Anhydrous (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
466085
Cerium(III) iodide,
AnhydroBeads
™
, −10 mesh, 99.99% trace metals basis
Expand
View Pricing