Castor Oil, Ethoxylated
Synonym(s):
Cremophor® EL, EL, Macrogolglycerol ricinoleate, PEG-35 castor oil, Polyoxyl-35 castor oil, Cremophor® ELP, ELP
CAS Number:
Products
Product Category
Brand
pH Value
Physical Form
Quality Segment
Application
Showing 1-2 of 2
Compare
Product Number
Description
Pricing