Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Carbon dioxide-
18
O
2
Linear Formula:
C
18
O
2
CAS Number:
2537-69-1
Molecular Weight:
48.01
Products
Product Category
building blocks (4)
stable isotopes (4)
gases (3)
Brand
Sigma-Aldrich (4)
Formula Weight
Melting Point (°C)
Isotope
Filter Products
Showing 1-4 of 4
Compare
Product Number
Description
Pricing
609714
Carbon dioxide-
18
O
2
,
97 atom %
18
O
Expand
View Pricing
609692
Carbon dioxide-
18
O
2
,
95 atom %
18
O
Expand
View Pricing
588601
Carbon dioxide-
18
O
2
,
50 atom %
18
O
Expand
View Pricing
609722
Carbon dioxide-
18
O
2
,
85 atom %
18
O
Expand
View Pricing