Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Calcium hydroxide
Synonym(s):
Calcium hydroxide
Linear Formula:
Ca(OH)
2
CAS Number:
1305-62-0
Molecular Weight:
74.09
Products
Product Category
bases (2)
inorganic bases (1)
salts (1)
Brand
SAFC (2)
Sigma-Aldrich (2)
Supelco (2)
Formula Weight
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Application
Reference Material Type
Special Grade
ACS reagent (1)
analytical standard (1)
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
239232
Calcium hydroxide,
ACS reagent, ≥95.0%
Expand
View Pricing
1.02047
Calcium hydroxide,
for analysis EMSURE
®
ACS,Reag. Ph Eur
Expand
View Pricing
450146
Calcium hydroxide,
99.995% trace metals basis
Expand
View Pricing
1.02119
Calcium hydroxide,
precipitated (≤ 0.0005% Al), EMPROVE
®
ESSENTIAL, USP, FCC, E 526
Expand
View Pricing
1.02110
Calcium hydroxide,
EMPROVE
®
ESSENTIAL, Ph. Eur., BP, JP, USP, FCC, E 526
Expand
View Pricing
1.02111
Calcium hydroxide,
for the preparation of DIN 19266 pH standard buffer solutions
Expand
View Pricing