Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Cadmium sulfate
8
/
3
-hydrate
Linear Formula:
CdSO
4
· 8/3H
2
O
CAS Number:
7790-84-3
Molecular Weight:
256.57
EC Number:
233-331-6
Products
Product Category
salts (1)
Brand
Sigma-Aldrich (1)
Formula Weight
Physical Form
Purity
Quality Segment
Special Grade
ACS reagent (1)
Puriss (1)
Puriss P.A. (1)
Filter Products
Showing 1-1 of 1
Compare
Product Number
Description
Pricing
20920
Cadmium sulfate
8
/
3
-hydrate,
puriss. p.a., ACS reagent, ≥99.0% (calc. based on CdSO
4
·
8
/
3
H
2
O, KT)
Expand
View Pricing