
Cadmium ionophore I

Synonym(s):
N,N,N′,N′-Tetrabutyl-3,6-dioxaoctanedi(thioamide), ETH 1062
Empirical Formula (Hill Notation):
C22H44N2O2S2
CAS Number:
Molecular Weight:
432.73
Products
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing