
Busulfan

Synonym(s):
1,4-Butanediol dimethanesulfonate, Busulfan
Linear Formula:
CH3SO2O(CH2)4OSO2CH3
CAS Number:
Molecular Weight:
246.30
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3