
Budesonide Related Compound G

Synonym(s):
16α,17-[Butylidenebis(oxy)]-11β, 21-dihydroxypregna-4-ene-3,20-dione, 1,2-Dihydrobudesonide
Empirical Formula (Hill Notation):
C25H36O6
CAS Number:
Molecular Weight:
432.55
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2