Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Bromocresol purple
Synonym(s):
Bromocresol purple, 5,5′-Dibromo-
o
-cresolsulfonphthalein, Bromcresol purple sultone form
Empirical Formula (Hill Notation):
C
21
H
16
Br
2
O
5
S
CAS Number:
115-40-2
Molecular Weight:
540.22
EC Number:
204-087-8
Products
Product Category
stains (1)
Brand
Supelco (1)
Color
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Agency
Chemical Composition
Quality Segment
Application
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
1.03025
Bromocresol purple,
pH indicator, ACS, Reag. Ph Eur
Expand
View Pricing
114375
Bromocresol purple,
BioReagent, suitable for indicator, Dye content 90 %
Expand
View Pricing
B5880
Bromocresol purple,
Powder
Expand
View Pricing