
Brexpiprazole-D8 solution

Empirical Formula (Hill Notation):
C25H19D8N3O2S
CAS Number:
Molecular Weight:
441.62
Products
Brand
Formula Weight
Manufacturer
Format
Quality Segment
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing