Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
Transferpettor positive displacement pipette, fixed volume
Products
Product Category
pipettors (9)
Manufacturer
Volume (µL)
Electronic
Package Size
Filter Products
Showing 1-9 of 9
Compare
Product Number
Description
Pricing
Z328340
BRAND
®
Transferpettor positive displacement pipette, fixed volume,
volume 200 μL, accuracy: 1.5%, precision: 0.2%
Expand
View Pricing
Z328324
BRAND
®
Transferpettor positive displacement pipette, fixed volume,
volume 50 μL, accuracy: 1.8%, precision: 0.4%
Expand
View Pricing
Z328235
BRAND
®
Transferpettor positive displacement pipette, fixed volume,
volume 1 μL, accuracy: 12%, precision: 4.0%
Expand
View Pricing
Z328332
BRAND
®
Transferpettor positive displacement pipette, fixed volume,
volume 100 μL, accuracy: 1.5%, precision: 0.4%
Expand
View Pricing
Z328308
BRAND
®
Transferpettor positive displacement pipette, fixed volume,
volume 20 μL, accuracy: 2.4%, precision: 0.5%
Expand
View Pricing
Z328294
BRAND
®
Transferpettor positive displacement pipette, fixed volume,
volume 10 μL, accuracy: 3.0%, precision: 0.8%
Expand
View Pricing
Z328243
BRAND
®
Transferpettor positive displacement pipette, fixed volume,
volume 2 μL, accuracy: 7.5%, precision: 2.0%
Expand
View Pricing
Z328286
BRAND
®
Transferpettor positive displacement pipette, fixed volume,
volume 5 μL, accuracy: 3.0%, precision: 0.8%
Expand
View Pricing
Z328316
BRAND
®
Transferpettor positive displacement pipette, fixed volume,
volume 25 μL, accuracy: 2.4%, precision: 0.4%
Expand
View Pricing