Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
BRAND
®
strips of 8 caps
Products
Product Category
caps (6)
Manufacturer
Color
Package Size
Technique
Filter Products
Showing 1-6 of 6
Compare
Product Number
Description
Pricing
BR781340
BRAND
®
strips of 8 caps,
polypropylene, clear, for PCR tubes
Expand
View Pricing
BR781334
BRAND
®
strips of 8 caps,
polypropylene, clear, flat
Expand
View Pricing
BR781344
BRAND
®
strips of 8 caps,
polypropylene, blue, for PCR tubes
Expand
View Pricing
BR781343
BRAND
®
strips of 8 caps,
polypropylene, green, for PCR tubes
Expand
View Pricing
BR781341
BRAND
®
strips of 8 caps,
polypropylene, rose, for PCR tubes
Expand
View Pricing
BR781342
BRAND
®
strips of 8 caps,
polypropylene, yellow, for PCR tubes
Expand
View Pricing