Skip to Content
AO
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Brady
®
thermal labeling system (TLS) printer accessories
Products
Manufacturer
Color
Filter Products
Showing 1-7 of 7
Compare
Product Number
Description
Pricing
Z663999
Brady
®
thermal labeling system (TLS) printer accessories,
ribbon R4310, black (wax-resin)
Expand
View Pricing
Z663972
Brady
®
thermal labeling system (TLS) printer accessories,
ribbon R6210, black (resin)
Expand
View Pricing
Z663212
Brady
®
thermal labeling system (TLS) printer accessories,
220V AC adapter for TLS2200
Expand
View Pricing
Z663220
Brady
®
thermal labeling system (TLS) printer accessories,
240 V AC adapter for TLS2200
Expand
View Pricing
Z663255
Brady
®
thermal labeling system (TLS) printer accessories,
hard sided carrying case for TLS2200
Expand
View Pricing
Z663263
Brady
®
thermal labeling system (TLS) printer accessories,
soft carrying case for TLS2200
Expand
View Pricing
Z663328
Brady
®
thermal labeling system (TLS) printer accessories,
communications cables for TLSPCLink
Expand
View Pricing