Borane dimethyl sulfide complex solution
Synonym(s):
BMS, Trihydro[thiobis[methane]]boron
Linear Formula:
(CH3)2S · BH3
CAS Number:
Molecular Weight:
75.97
Products
Brand
Formula Weight
Physical Form
Quality Segment
Functional Group
Markush Class
Markush Group
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing