Skip to Content
Merck
Boc-OSu ≥98.0% (CHN)

Boc-OSu

Boc-OSu ≥98.0% (CHN)
Synonym(s):
N-(tert-Butoxycarbonyloxy)succinimide, tert-Butyl N-succinimidyl carbonate
Empirical Formula (Hill Notation):
C9H13NO5
CAS Number:
Molecular Weight:
215.20
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Application
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing