
Bismuth(III) trifluoromethanesulfonate

Synonym(s):
Bi(OTf)3, Bismuth tris(trifluoromethanesulfonate), Bismuth(III) triflate
Linear Formula:
Bi(OSO2CF3)3
CAS Number:
Molecular Weight:
656.19
Products
Brand
Formula Weight
Melting Point (°C)
Markush Class
Markush Group
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing