
Bismuth citrate

Synonym(s):
Bismuth(III) citrate, Bismuth citrate
Linear Formula:
[O2CCH2C(OH)(CO2)CH2CO2]Bi
CAS Number:
Molecular Weight:
398.08
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing