
Bis(ethylcyclopentadienyl)cobalt(II)

Synonym(s):
(EtCp)2Co, 1,1′-Diethylcobaltocene
Empirical Formula (Hill Notation):
C14H18Co
CAS Number:
Molecular Weight:
245.23
Products
Brand
Formula Weight
Physical Form
Quality Segment
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing