Skip to Content
Merck
Biphenyl Pharmaceutical Secondary Standard; Certified Reference Material

Biphenyl

Biphenyl Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Biphenyl
Linear Formula:
C6H5C6H5
CAS Number:
Molecular Weight:
154.21

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Package Size
API Family
Reference Material Type
Special Grade
Technique