
Bifenox

Synonym(s):
Methyl 5-(2,4-dichlorophenoxy)-2-nitrobenzoate
Empirical Formula (Hill Notation):
C14H9Cl2NO5
CAS Number:
Molecular Weight:
342.13
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2