Skip to Content
Merck
Bicinchoninic acid disodium salt hydrate ≥98% (HPLC)

Bicinchoninic acid disodium salt hydrate

Bicinchoninic acid disodium salt hydrate ≥98% (HPLC)
Synonym(s):
2,2′-Biquinoline-4,4′-dicarboxylic acid disodium salt, 4,4′-Dicarboxy-2,2′-biquinoline disodium salt, BCA, Disodium 2,2′-biquinoline-4,4′-dicarboxylate
Empirical Formula (Hill Notation):
C20H10N2Na2O4 · xH2O
CAS Number:
Molecular Weight:
388.28 (anhydrous basis)

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Application
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing