Skip to Content
Merck
Betahistine dihydrochloride European Pharmacopoeia (EP) Reference Standard

Betahistine dihydrochloride

Betahistine dihydrochloride European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Methyl[2-(2-pyridyl)ethyl]amine dihydrochloride
Empirical Formula (Hill Notation):
C8H12N2 · 2HCl
CAS Number:
Molecular Weight:
209.12

Products

Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
API Family
Special Grade