Benzyl 2,2,2-trichloroacetimidate
Synonym(s):
2,2,2-Trichloroacetimidic acid benzyl ester
Linear Formula:
CCl3C(=NH)OCH2C6H5
CAS Number:
Molecular Weight:
252.52
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1